Order Entry
Austria
ContactUsLinkComponent
Ethyl acetate ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Flammable
Irritant
Ethyl acetate ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Catalog # 85481.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Ethyl acetate ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Catalog # 85481.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance:
    Clear colorless liquid
  • Acidity (as Acetic acid):
    max. 0.003 %
  • Alkalinity (as Ammonia):
    max. 0.0005 %
  • Color (APHA, Pt-Co):
    max. 10
  • Assay (GC, on anhydrous basis):
    min. 99.9 %
  • Residue after evaporation:
    max. 0.0005 %
  • Water (KF):
    max. 0.05 %
  • T260nm:
    min. 75 %
  • T275nm:
    min. 98 %
  • LC-MS suitability test:
    Complies
  • Ag (Silver):
    max. 0.1 ppm
  • Al (Aluminum):
    max. 0.05 ppm
  • Ba (Barium):
    max. 0.1 ppm
  • Bi (Bismuth):
    max. 0.1 ppm
  • Ca (Calcium):
    max. 0.05 ppm
  • Cd (Cadmium):
    max. 0.05 ppm
  • Co (Cobalt):
    max. 0.05 ppm
  • Cr (Chromium):
    max. 0.02 ppm
  • Cu (Copper):
    max. 0.02 ppm
  • Fe (Iron):
    max. 0.02 ppm
  • K (Potassium):
    max. 0.05 ppm
  • Li (Lithium):
    max. 0.1 ppm
  • Mg (Magnesium):
    max. 0.05 ppm
  • Mn (Manganese):
    max. 0.02 ppm
  • Mo (Molybdenum):
    max. 0.05 ppm
  • Na (Sodium):
    max. 0.05 ppm
  • Ni (Nickel):
    max. 0.02 ppm
  • Pb (Lead):
    max. 0.02 ppm
  • Sn (Tin):
    max. 0.05 ppm
  • Sr (Strontium):
    max. 0.05 ppm
  • Zn (Zinc):
    max. 0.1 ppm

Specification Test Results

About this item

 :
 :88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119475103-46
 :GHS02
 :GHS07
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171