Order Entry
Austria
Orders LinkContactUsLinkComponent
Microtubes, 1,1 ml

Microtubes, 1,1 ml

In-line caps for 1,1 ml microtubes
Catalog # PIER15088
Microtubes, 1,1 ml
Catalog # PIER15088
Microtubes, 1,1 ml
Microtubes, 1,1 ml

In-line caps for 1,1 ml microtubes

Price
65,50 €
Log in for availability
1 * 120 ST
Product Details & Documents

Microtube racke system consists of 1,1 ml-tall microtubes supplied pre-racked in boxes to match a standard 96-well microplate footprint.

  • Available individually or in an 8×12 microtitration plate format
  • 8×12 format simplifies and speeds applications in which a multi-channel pipettor is used
  • Format is compatible with robotic workstations
  • Microtube system can be centrifuged

The microtubes resemble 1 ml pipette tips, except that they have round bottoms. Like a box of 1 ml pipette tips, the racked microtubes match multi-channel pipettors, providing convenience for a variety of reagent- and sample-dispensing procedures.

Specifications
  • Description:
    In-line caps for 1,1 ml microtubes
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.