ContactUsLinkComponent
VWR® Hitachi Chromaster HPLC Control Interfaces for HPLC System
VWR® Hitachi Chromaster HPLC Control Interfaces for HPLC System
Catalog # 903-0545
Supplier:  VWR Collection
undefined
VWR® Hitachi Chromaster HPLC Control Interfaces for HPLC System
Catalog # 903-0545
Supplier:  VWR Collection
Specifications
  • Description:
    Chromaster Plus HPLC interface control board for Chromaster Plus system control. USB cable and e-line cable (0,5 m) included, built into the autosampler upon installation in laboratory, if no autosampler is used, then the interface box (S) must be ordered instead
  • Seq:
    44
  • Pk:
    1