Reduced-serum media, Hyclone™ HyQ-RS
Supplier: HyClone products (Cytiva)
Certificates
About this item
Hyclone™ HyQ-RS is a reduced-serum (RS) media for applications that require a lower concentration of serum supplementation.
- Reduce FBS supplementation rate to 2 to 4% for most cell culture applications
- Suitable for a wide variety of cell types normally used with standard classical formulations
- Maintain cell performance while reducing serum supplementation by up to 80%
- All contain phenol red
- 0,1 μm sterile filtered
- Manufactured using ISO 9001:2000 certified processes
We can determine which (if any) ingredients in a medium formulation are of animal origin or, if they have come in contact with animal derived materials in manufacture.
High yield cultures begin with high quality media. In order to maintain these standards, all HyClone products as well as rea material components must pass strict Quality Control testing to confirm the appropriate level of quality. Each raw material has a specification document describing the product, the required grade or purity, the source, and the criteria for acceptance on incoming inspection.
Frequently Bought Together