Order Entry
Austria
Orders LinkContactUsLinkComponent
Blocking buffer, concentrate, Pierce™ Clear Milk

Blocking buffer, concentrate, Pierce™ Clear Milk

Pierce™ Clear Milk blocking buffer (10X), 100 mL
Catalog # PIER37587
Blocking buffer, concentrate, Pierce™ Clear Milk
Catalog # PIER37587
CAS Number:  
Blocking buffer, concentrate, Pierce™ Clear Milk
Blocking buffer, concentrate, Pierce™ Clear Milk

Pierce™ Clear Milk blocking buffer (10X), 100 mL

CAS Number:  
Price
231,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

This clear milk blocking buffer is a long shelf life, high performance replacement for homemade milk blocking buffers in Western blotting applications.

  • Kit stable for 1 year stored at 4 °C
  • Convenient and saves storage space
  • Easy to use with ready-to-dilute solution

It is a proprietary pre-formulated milk solution for blocking excess non specific binding sites, reducing background in Western blotting applications and diluting antibodies when used with nitrocellulose and PVDF membranes. The solution provides lower background, enhanced sensitivity, extended shelf life and reproducible results compared to homemade buffers based on dissolved non fat dry milk. For Research Use Only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Pierce™ Clear Milk blocking buffer (10X)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.