Order Entry
Austria
Orders LinkContactUsLinkComponent
Blocker™ Blocking Buffer, Casein

Blocker™ Blocking Buffer, Casein

Blocker™ Blocking Buffer, Casein
Certificates
Blocker™ Blocking Buffer, Casein

3 Options Available Below

Product Details & Documents

Blocker™ Casein in PBS or TBS are ready-to-use, solutions of purified casein protein for blocking steps in Western blot, ELISA, IHC and nucleic acid detection methods.

  • Easy to use
  • Safe, stable and thimerosal-free formulations
  • Purified casein provides fewer chances of cross reaction
  • Flexible and available in PBS and TBS formulations

This blocking buffer contains casein protein that is purified from milk by the Hammarsten method. The phosphate and Tris-buffered saline formulations are used for blocking excess binding sites on membranes and microplates in antibody based detection methods. Blocker Casein buffers are 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • PIER37528
  • PIER37582
  • PIER37583
  • Concentration:
  • 1X
  • 1X
  • 1X
  • For use with:
  • Western blot, ELISA
  • Western blot, ELISA
  • Western blot, ELISA
  • Physical form:
  • Liquid
  • Liquid
  • Liquid
  • Storage buffer:
  • PBS
  • PBS
  • TBS

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • PIER37528
    Blocker™ Casein in PBS, blocking buffer
    Blocker™ Casein in PBS, blocking buffer
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 230,00 €
    230,00 €
    1 * 1 L
       230,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37528
    Concentration
    1X
    For use with
    Western blot, ELISA
    Physical form
    Liquid
    Storage buffer
    PBS
  • PIER37582
    Blocker™ Casein in PBS, blocking buffer
    Blocker™ Casein in PBS, blocking buffer
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 57,50 €
    57,50 €
    1 * 100 mL
       57,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37582
    Concentration
    1X
    For use with
    Western blot, ELISA
    Physical form
    Liquid
    Storage buffer
    PBS
  • PIER37583
    Blocker™ Casein in TBS, blocking buffer
    Blocker™ Casein in TBS, blocking buffer
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 57,00 €
    57,00 €
    1 * 100 mL
       57,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37583
    Concentration
    1X
    For use with
    Western blot, ELISA
    Physical form
    Liquid
    Storage buffer
    TBS
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.