Order Entry
Austria
Orders LinkContactUsLinkComponent
StartingBlock™ Blocking Buffer

StartingBlock™ Blocking Buffer

StartingBlock™ Blocking Buffer
Certificates
StartingBlock™ Blocking Buffer

6 Options Available Below

Product Details & Documents

These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.

  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS
  • For research use only
  • Not for use in diagnostic procedures

It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock blocking buffer is available in PBS and TBS, with and without Tween 20.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • PIER37578
  • PIER37579
  • PIER37538
  • PIER37539
  • PIER37542
  • PIER37543
  • Concentration:
  • 1X
  • 1X
  • 1X
  • 1X
  • 1X
  • 1X
  • For use with:
  • Western blot, ELISA, Immunohistochemistry (IHC)
  • Western blot, ELISA, Immunohistochemistry (IHC)
  • Western blot, ELISA, Immunohistochemistry (IHC)
  • Western blot, ELISA
  • Western blot, ELISA, Immunohistochemistry (IHC)
  • Western blot, ELISA
  • Physical form:
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Storage buffer:
  • PBS
  • TBS
  • PBS
  • PBST
  • TBS
  • TBST

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • PIER37578
    StartingBlock™ (PBS), blocking buffer, 100 ml
    StartingBlock™ (PBS), blocking buffer, 100 ml
    100 ml
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 64,30 €
    64,30 €
    1 * 100 mL
       64,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37578
    Concentration
    1X
    For use with
    Western blot, ELISA, Immunohistochemistry (IHC)
    Physical form
    Liquid
    Storage buffer
    PBS
  • PIER37579
    StartingBlock™ (TBS), blocking buffer, 100 ml
    StartingBlock™ (TBS), blocking buffer, 100 ml
    100 ml
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 64,30 €
    64,30 €
    1 * 100 mL
       64,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37579
    Concentration
    1X
    For use with
    Western blot, ELISA, Immunohistochemistry (IHC)
    Physical form
    Liquid
    Storage buffer
    TBS
  • PIER37538
    StartingBlock™ (PBS), blocking buffer, 1 L
    StartingBlock™ (PBS), blocking buffer, 1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 349,00 €
    349,00 €
    1 * 1 L
       349,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37538
    Concentration
    1X
    For use with
    Western blot, ELISA, Immunohistochemistry (IHC)
    Physical form
    Liquid
    Storage buffer
    PBS
  • PIER37539
    StartingBlock™ T20 (PBS), blocking buffer with 0,05% Tween®20 detergent, 1 L
    StartingBlock™ T20 (PBS), blocking buffer with 0,05% Tween®20 detergent, 1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 371,00 €
    371,00 €
    1 * 1 L
       371,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37539
    Concentration
    1X
    For use with
    Western blot, ELISA
    Physical form
    Liquid
    Storage buffer
    PBST
  • PIER37542
    StartingBlock™ (TBS), blocking buffer, 1 L
    StartingBlock™ (TBS), blocking buffer, 1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37542
    Concentration
    1X
    For use with
    Western blot, ELISA, Immunohistochemistry (IHC)
    Physical form
    Liquid
    Storage buffer
    TBS
  • PIER37543
    StartingBlock™ T20 (TBS), blocking buffer with 0,05% Tween®20 detergent, 1 L
    StartingBlock™ T20 (TBS), blocking buffer with 0,05% Tween®20 detergent, 1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 371,00 €
    371,00 €
    1 * 1 L
       371,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    PIER37543
    Concentration
    1X
    For use with
    Western blot, ELISA
    Physical form
    Liquid
    Storage buffer
    TBST
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.