Western blot enhancer, SuperSignal™, Pierce
Supplier: Thermo Fisher Scientific
Certificates
About this item
SuperSignal™ Western Blot Enhancer is a membrane treatment and a primary antibody diluent that has been shown to increase intensity of a target protein band on a Western blot and to enable detection of a target analyte at a level at which it could not be detected without enhancement.
- Increase in signal intensity and sensitivity
- Improves signal-to-noise ratio for poor quality and low affinity antibodies
- Provides effective signal enhancement with PVDF and nitrocellulose membranes
- Ready-to-use and can be stored at room temperature
Frequently Bought Together