Order Entry
Austria
ContactUsLinkComponent
Recombinant protein L, biotinylated, Pierce™
Recombinant protein L, biotinylated, Pierce™
  PIER29997
 :  
undefined
Recombinant protein L, biotinylated, Pierce™
  PIER29997
 :  

 

  • Pk:
    0,5 mg
  • Description:
    Recombinant Protein L, biotinylated

 

 

Pierce™ Biotinylated Recombinant Protein L can be used to probe and detect mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.

  • Contains four immunoglobulin binding domains per protein
  • Binds members of all classes of Ig that contain kappa light chains
  • Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen binding site.