Penicillin/Streptomycin/Fungizone solution 100X, HyClone™
Artikelnr. HYCLSV30079.01
Supplier: HyClone products (Cytiva)
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Format:Liquid
- Storage:Store frozen
- Antibiotic Spectrum:Broad spectrum antibacterial and antifungal
- Concentration:100X
- Shelf life:18 months
- Sterilization:0.2 µm sterile filtered
Specification Test Results
About this item
: