ContactUsLinkComponent
Pierce™ Agarose ChIP Kit
Pierce™ Agarose ChIP Kit
Catalog # PIER26156
CAS Number:  
undefined
Pierce™ Agarose ChIP Kit
Catalog # PIER26156
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Agarose ChIP kit, sufficient to perform ChIP reactions for 30 target proteins from 10 sample conditions
Specifications
About this item
Provides a complete set of reagents and a simple, fast and reproducible protocol to perform chromatin immunoprecipitation (ChIP) assays.
  • High-recovery DNA purification
  • Highly efficient isolation and lysis of nuclei
  • Easy and reproducible enzymatic digestion
  • Low-background and high-binding capacity Protein A/G agarose resin
  • Highly specific positive controls included: RNA polymerase II antibody and GAPDH PCR primers
ChIP assay has the ability to capture a snapshot of specific protein-DNA interactions as they occur in living cells, and then quantitate the interactions using standard or quantitive PCR.