Orders LinkContactUsLinkComponent
Visual colour kits, oxygen, MQuant®, Supelco® | Avantor
Visual colour kits, oxygen, MQuant®, Supelco®
Supplier:   Merck
Visual colour kits, oxygen, MQuant®, Supelco®
Supplier:  Merck
Certificates
Visual colour kits, oxygen, MQuant®, Supelco®
2 Options Available Below
Product Details & Documents
Quick tests, visual colour comparison.
  • Results in approximately 2 minutes
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
MQuant® titrimetric and colorimetric methods:
This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.
Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1.11107.0001
  • 1.11152.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • Range
    Chemical method
    Type
    Pk
    Availability
    Price
  • Oxygen Test
    0,1-10 mg/l O₂
    Winkler (modified)
    MTC
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 218,00 €
    218,00 €
    1 * 100 Tests
       218,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.11107.0001
    M-Clarity quality level
    MQ100
  • 1.11152.0001
    Oxygen test, RP for 1.11107.0001
    Oxygen test, RP for 1.11107.0001
    Winkler (modified)
    MTC
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 190,00 €
    190,00 €
    1 * 100 Tests
       190,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.11152.0001
    M-Clarity quality level
    MQ100
Accessories
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Oxygen reaction tube for 1.14662.0001 (MTC)
    Oxygen reaction tube for 1.14662.0001 (MTC)
    1.14663.0001
    MTC
    Log in for availability
    1 * 1 ST - 97,40 €