Orders LinkContactUsLinkComponent
Visual colour kits, chloride, MQuant®, Supelco®
Visual colour kits, chloride, MQuant®, Supelco®
Visual colour kits, chloride, MQuant®, Supelco®
Supplier:  Merck
Certificates
Visual colour kits, chloride, MQuant®, Supelco®
Visual colour kits, chloride, MQuant®, Supelco®
3 Options Available Below
Product Details & Documents
Quick tests, visual colour comparison.
  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
MQuant® with colour card comparator
By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/L). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
MQuant® with colour disc comparator MCD
Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.
Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart or comparator wheel and extensive instructions for use.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1.14753.0001
  • 1.18322.0002
  • 1.14401.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • Range
    Chemical method
    Type
    Pk
    Availability
    Price
  • Chloride test
    3-6-10-18-30-60-100-180-300 mg/l Cl
    Mercury (II) thiocyanate
    MCD
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 301,00 €
    301,00 €
    1 * 200 Tests
       301,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14753.0001
    M-Clarity quality level
    MQ100
  • 1.18322.0002
    Chloride test, RP for 1.14753.0001; 1.14401.0001
    Chloride test, RP for 1.14753.0001; 1.14401.0001
    Mercury (II) thiocyanate
    MCD
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 165,00 €
    165,00 €
    1 * 200 Tests
       165,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.18322.0002
    M-Clarity quality level
    MQ100
  • Chloride test
    5-10-20-40-75-150-300 mg/l Cl
    Mercury (II) thiocyanate
    MCC
    400 Tests
    Log in for availability
    Log in for availability
    1 * 400 Tests
    1 * 400 Tests - 174,00 €
    174,00 €
    1 * 400 Tests
       174,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14401.0001
    M-Clarity quality level