ContactUsLinkComponent
Biohazard disposal can, SP Bel-Art
Biohazard disposal can, SP Bel-Art
Catalog # 817-0453
undefined
Biohazard disposal can, SP Bel-Art
Catalog # 817-0453
Specifications
  • Colour:
    Red
  • Material:
    PP
  • W×D×H:
    225×305×290 mm
  • Pk:
    1
Specifications
About this item
Biohazard disposal can with lift-up cover.
  • One piece hinged lid protects users from contact with contents; helps retain odours and contain airborne contaminants
  • Moulded out of polypropylene for long service life
  • Puncture resistant, autoclavable at 121 °C (250 °F)
  • 482,6×584,2 mm autoclave bags fit perfectly and are secured with an internal stainless steel wire bar
Suitable for biological/biohazard materials, broken glass, recyclables or other waste and sized for benchtop or floor placement.