Cell culture media for SILAC
Supplier: Thermo Fisher Scientific
Certificates
About this item
Thermo Scientific™ cell culture media for SILAC is optimised for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyse protein expression by mass spectrometry (MS).
- Sterile, endotoxin-free and cell culture-compatible
- Flexible with liquid media deficient in both L-lysine and L-arginine
Allows more complete proteome coverage through multiple isotopic amino acid labeling. This is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together