Order Entry
Austria
ContactUsLinkComponent
 
Universal nuclease for cell lysis, Pierce™
 
undefined
Universal nuclease for cell lysis, Pierce™

 

  • Storage conditions:
    Store in a cool, dry, well-ventilated area, protected from direct sunlight
  • Purity:
    99% enzyme
  • Concentration:
    250 U/µl

 

Frequently Bought Together