Order Entry
Austria
ContactUsLinkComponent
 
E-Z 96® Plant DNA DS kit
 
E-Z 96® Plant DNA DS kit

 

  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Sample size:
    50 mg of wet tissue or 10 mg dry tissue
  • Description:
    E-Z 96® Plant DNA DS Kit

 

Frequently Bought Together