ContactUsLinkComponent
Pierce™ Alkaline Phosphatase, Biotinylated
Pierce™ Alkaline Phosphatase, Biotinylated
Catalog # PIER29339
CAS Number:  
undefined
Pierce™ Alkaline Phosphatase, Biotinylated
Catalog # PIER29339
CAS Number:  
Specifications
  • Pk:
    1 mg
  • Description:
    Alkaline phosphatase, biotinylated
Specifications
About this item
Biotinylated alkaline phosphatase is most commonly used in immunohistochemistry (IHC) to amplify the signal of biotinylated primary antibodies using the ABC staining method.