Order Entry
Austria
ContactUsLinkComponent
Dry buffer packs, Carbonate-Bicarbonate Buffer, JAW™
Dry buffer packs, Carbonate-Bicarbonate Buffer, JAW™
Catalog # 786-291
Supplier:  G-Biosciences
CAS Number:  
Dry buffer packs, Carbonate-Bicarbonate Buffer, JAW™
Catalog # 786-291
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    20
  • Description:
    Carbonate-bicarbonate buffer packs, JAW™, 1X, 1 l/pack
  • Seq:
    01
  • pH value:
    9,4
  • Contents:
    0,2 M Carbonate-bicarbonate buffer

Specifications

About this item

Premixed buffer powders that simply require adding deionised water to generate a ready-to-use buffer.

JAW™ Carbonate-Bicarbonate Buffer packs each make one litre of: 0,2M carbonate-bicarbonate buffer (at a pH of 9,4).