ContactUsLinkComponent
Pierce™ Polystyrene Strip Plates, 8-Well, Corner Notch
Pierce™ Polystyrene Strip Plates, 8-Well, Corner Notch
Catalog # PIER15031
undefined
Pierce™ Polystyrene Strip Plates, 8-Well, Corner Notch
Catalog # PIER15031
Specifications
  • Colour:
    Clear
  • Description:
    96-well strip microplate assembled in frame, 1×8 per strip, flat bottom, corner notch
  • Pk:
    100
  • Well volume:
    360 µl
Specifications
About this item
PS, high antibody-binding 96-well microplate with individual 8-well strips in the plate frame for immunoassay applications.
  • Clear polystyrene for colorimetric detections
  • Plates are certified for performance, having consistently low coefficients of variation among wells
  • Hydrophobic-ionic binding interaction
  • Wells can be easily separated from strips with gentle twist
  • Corner notch for use in standard plate readers