Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid solution 6N, Sequencing grade, Pierce™
Corrosive
Toxic
Hydrochloric acid solution 6N, Sequencing grade, Pierce™
Catalog # PIER24308
CAS Number:  
undefined
Corrosive
Toxic
Hydrochloric acid solution 6N, Sequencing grade, Pierce™
Catalog # PIER24308
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1 SET
  • Seq:
    0125
  • Pk Info:
    10 x 1 ml

Specifications

About this item

 :
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS05
 :GHS06
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324