SMART Digest™ Chymotrypsin Kits
Supplier: Thermo Fisher Scientific
Certificates
About this item
SMART Digest™ Chymotrypsin Kits are designed to provide a complimentary digestion option to the SMART Digest™ Trypsin Kits which incorporate trypsin functionality. The Chymotrypsin enzyme functionality allows digestion of proteins at different points of the amino acid chain compared to trypsin. This allows for different/additional structural information to be obtained when trying to characterise a protein. This option can also be used if a protein does not contain the specific point where trypsin cuts the protein chain.
- Significantly faster than in-solution digestion
- Simple process
- High reproducibility
- Amenable to high-throughput processing
Delivery: Available as a stand alone kit, or with optional filter plate or SOLAµ™ SPE plate. Bulk resin (-B) and magnetic bulk resin (-MB) are also available separately.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together