ContactUsLinkComponent
Hardness tester
Hardness tester
Catalog # 649-0048
Supplier:  Industrial Physics
undefined
Hardness tester
Catalog # 649-0048
Supplier:  Industrial Physics
Specifications
  • Description:
    Buchholz hardness test
  • Pk:
    1
Specifications
About this item
The Buchholz hardness indentation test provides a method for an indentation test for single coating or multicoat system of paint, varnish, or related products.
  • Pins and cutting part in hardened steel preventing wear
  • Dual timer preset for both “in-position” and “recovering” time
  • Separate level gauge allows for checking the level of the test panel
Equipped with functional grips allowing gentle placing and lifting of the instrument. High quality microscope with precision optical lenses and strong illuminator for a clear visibility of the indentation mark. The calibrated weight with sharp-edged wheel is positioned on the test specimen for a set period of time. The length of the indentation mark in the coating indicates of the hardness of the surface.
Certifications: ISO 2815:2003
Delivery: Supplied with calibrated weight and metal wheel, illuminated microscope, level gauge, digital timer and two markers.