Order Entry
Austria
ContactUsLinkComponent
 
[Arg8]-Vasopressin, MP Biomedicals
 
[Arg8]-Vasopressin, MP Biomedicals

 

  • CAS No.:
    113-79-1
  • Storage conditions:
    −20 °C
  • Protein/peptide name:
    [Arg8]-Vasopressin
  • Purity:
    >95%
  • Colour:
    White
  • Formulation:
    Lyophilised powder

 

Frequently Bought Together