Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Daunosamine hydrochloride
Daunosamine hydrochloride
Daunosamine hydrochloride
Daunosamine hydrochloride
Artikelnr. ICNA0215227325
Supplier:
MP Biomedicals
CAS Number:
Expand image
Daunosamine hydrochloride
Artikelnr. ICNA0215227325
Supplier:
MP Biomedicals
CAS Number:
Specifications
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
:
197.659 g/mol
CAS Number:
32385-06-1
Formula:
C₇H₁₆ClNO₃
MDL Number:
MFCD00079420
Product Details