Order Entry
Austria
ContactUsLinkComponent
Leuconostoc mesenteroides Glucose-6-Phosphate dehydrogenase, MP Biomedicals
Leuconostoc mesenteroides Glucose-6-Phosphate dehydrogenase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Leuconostoc mesenteroides Glucose-6-Phosphate dehydrogenase, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9001-40-5
  • Species:
    Leuconostoc mesenteroides
  • Storage conditions:
    −20 °C
  • Form:
    White lyophilized powder
  • Enzyme name:
    Glucose-6-Phosphate Dehydrogenase
  • Enzyme activity:
    ≥400 u/mg solid
  • Solubility:
    Dissolves in 0,1 M TEA/NaOH pH 8,0 (5 mg/ ml-clear, colorless solution), also in distilled water or dilute buffer
  • Molecular weight:
    104 kDa
  • pH:
    5,5 - 7,5

Specifications

Frequently Bought Together