Order Entry
Austria
ContactUsLinkComponent
Test kits, complete boiler and cooling water, drop count titration
Test kits, complete boiler and cooling water, drop count titration
Catalog # HACH2350300
Supplier:  Hach
CAS Number:  
undefined
Test kits, complete boiler and cooling water, drop count titration
Catalog # HACH2350300
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Boiler feed and scale test kit, BSC-1
  • Seq:
    01

Specifications

About this item

This kit is used for controlling boiler and cooling water treatment and checking corrosion using the drop count test which is a simple volumetric/ titration method with colour indication. It is an economical solution for the operation of modern boilers and cooling towers.

  • Simple, accurate methods
  • All-inclusive, the reagents are supplied with the kit
  • Application specific