Bulb for pipettes, SP Bel-Art
Catalog # BELAH378880000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Safety bulb for pipettes
- Pk:1
Specifications
About this item
Pipette bulb made of silicone rubber.
- Tapered, silicone rubber seal provides an airtight fit on all size pipettes
- Can be opened, cleaned and steam autoclaved at 121 °C
- Provided with an elastic cord for dedicating that pipettor to a specific reagent bottle
- For use with Westergren or Wintrobe sedimentation tubes