Order Entry
Austria
ContactUsLinkComponent
Warming plate, 37 °C, Bel-Art ProCulture, SP Bel-Art
Warming plate, 37 °C, Bel-Art ProCulture, SP Bel-Art
Catalog # BELAF370151000
undefined
Warming plate, 37 °C, Bel-Art ProCulture, SP Bel-Art
Catalog # BELAF370151000

Specifications

  • Temp.:
    37 °C
  • W×D×H:
    249×254×57 mm
  • Pk:
    1
  • Plate size:
    178×203 mm

Specifications

About this item

Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37 °C (±1 °C) outside of the incubator.

  • Reduces cell stress and allows enzymes like trypsin to work efficiently
  • Can be used to maintain the warmth of reagents inside the hood
  • Fits easily in cell culture hoods or on the workbench
  • Temperature remains uniform across the entire anodized aluminum warming plate
  • 15-minute count down timer keeps track of time sensitive applications
  • UV resistant polypropylene housing will not break down under germicidal UV lights
  • Water resistant, safe for cleaning with ethanol or IPA

Avoids numerous entries into the CO₂ incubator which alter its atmosphere, harming ongoing experiments in the incubated cultures.

Certifications: 220 V is CE marked.