Orders LinkContactUsLinkComponent
Settlometer kit
Settlometer kit
Settlometer kit
Catalog # HACH2115700
Supplier:   Hach
Settlometer kit
Catalog # HACH2115700
Supplier:  Hach
Settlometer kit
Settlometer kit
Settlometer kit
Price
336,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
This kit is used for analyzing activated sludge process. Custom PP paddle allows agitation of sample without scratching the jar.
  • Easy-to-read white graduations
  • Impact-resistant
  • Chemically resistant at pH range 3 to 10
Delivery information: Kit includes a transparent PC 2 liter jar with cover and 10 PC graduated centrifuge tubes (15 ml).
Specifications
  • Description:
    Settlometer kit