Vitamin A acetate USP
Catalog # SPCMV1257-25KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Fiber drum
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:328.49 g/mol
EINECS:204-844-2
CAS Number:127-47-9
Formula:C₂₂H₃₂O₂
MDL Number:MFCD00019413