Order Entry
Austria
Orders LinkContactUsLinkComponent
Sorbitan monooleate NF

Sorbitan monooleate NF

Sorbitan monooleate NF
Supplier:  Spectrum Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Sorbitan oleate, Span 80, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol mono-(9Z)-9-octadecenoate
  • CAS Number:1338-43-8
  • Formula:C₂₄H₄₄O₆
  • MDL Number:MFCD00080948
  • Molecular Weight:428.61 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-665-4

Documents

Specifications for Product Family

Specification Test Results

  • Assay:
    72% Min.
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.