Order Entry
Austria
Orders LinkContactUsLinkComponent
L(+)-Histidine USP

L(+)-Histidine USP

Supplier:   Spectrum Chemical
L(+)-Histidine USP
Supplier:  Spectrum Chemical
Certificates
L(+)-Histidine USP

2 Options Available Below

Product Details & Documents

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Melting Pt:272…273 °C
  • Density:1.44 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-745-3
Specifications for Product Family

Specification Test Results

  • Assay:
    98.5% Min.
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 131,00 €
    131,00 €
    1 * 25 g
       131,00 €

  • Poly bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1 610,00 €
    1 610,00 €
    1 * 1 kg
       1 610,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.