Fumaric acid NF
Supplier: Spectrum Chemical
About this item
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: trans-1,2-Ethylentricarboxylic acid
- CAS Number:110-17-8
- Formula:C₄H₄O₄
- MDL Number:MFCD00002700
- Molecular Weight:116.07 g/mol
- Merck Index:13,04308
- Boiling Pt:356 °C (1013 hPa)
- Melting Pt:287 °C
- Density:1.625 g/cm³ (20 °C)
- Flash Pt:273 °C
- EINECS:203-743-0
3 Options Available Below
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: trans-1,2-Ethylentricarboxylic acid
- CAS Number:110-17-8
- Formula:C₄H₄O₄
- MDL Number:MFCD00002700
- Molecular Weight:116.07 g/mol
- Merck Index:13,04308
- Boiling Pt:356 °C (1013 hPa)
- Melting Pt:287 °C
- Density:1.625 g/cm³ (20 °C)
- Flash Pt:273 °C
- EINECS:203-743-0
Specifications for Product Family
Specification Test Results
- Assay:99.5% Min.
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



