Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Acetanilide

Irritant
Supplier:   Spectrum Chemical
Acetanilide
Supplier:  Spectrum Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  N-Phenylacetamide
  • CAS Number:103-84-4
  • Formula:C₈H₉NO
  • MDL Number:MFCD00008674
  • Molecular Weight:135.17 g/mol
  • Merck Index:13,00051
  • Boiling Pt:304 °C (1013 hPa)
  • Melting Pt:115 °C
  • Density:1.22 g/cm³ (20 °C)
  • Flash Pt:174 °C
  • EINECS:203-150-7
Specifications for Product Family

Specification Test Results

  • Assay:
    99% Min.
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 466,00 €
    466,00 €
    1 * 1 kg
       466,00 €

  • Poly pail
    12 kg
    Log in for availability
    Log in for availability
    1 * 12 kg
    1 * 12 kg - 1 470,00 €
    1 470,00 €
    1 * 12 kg
       1 470,00 €

  • Fiber drum
    50 kg
    Log in for availability
    Log in for availability
    1 * 50 kg
    1 * 50 kg