Orders LinkContactUsLinkComponent
Supelco® Spectroquant® Test Kits, Copper Cell Test
Supelco® Spectroquant® Test Kits, Copper Cell Test
Copper cell test, 0,05 - 8,00 mg/l Cu, Cuprizone, Prove, Nova 30/60, Move, 25 Tests
Catalog # 1.14553.0001
Supplier:   Merck
Supelco® Spectroquant® Test Kits, Copper Cell Test
Catalog # 1.14553.0001
Supplier:  Merck
CAS Number:  
Supelco® Spectroquant® Test Kits, Copper Cell Test
Supelco® Spectroquant® Test Kits, Copper Cell Test
Copper cell test, 0,05 - 8,00 mg/l Cu, Cuprizone, Prove, Nova 30/60, Move, 25 Tests
CAS Number:  
Price
160,00 €
Log in for availability
1 * 25 Tests
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60 A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
For the determination of the total content of copper, please use the Crack Set (1.14688.0001) before the photometric procedure.
Specifications
Specifications
  • Chemical method:
    Cuprizone
  • Pk:
    25 Tests
  • Description:
    Copper cell test
  • For unit:
    Prove, Nova 30/60, Move
  • Range:
    0,05 - 8,00 mg/l Cu
  • Cat. No.:
    1.14553.0001
  • M-Clarity quality level:
    MQ100