
(1S,2S)-(+)-2-Benzyloxycyclohexylamine ≥99%, ee 99%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:216394-07-9
:
Formula:C₁₃H₁₉NO
MDL Number:MFCD01075753
:205.30 g/mol
Boiling Pt:79…80 °C (0.08 mmHg)
Density:1.02
Flash Pt:>110 °C (230 °F)
Storage Temperature:Refrigerator
UN:2735
ADR:8,III
:GHS05
Related Information
Frequently Bought Together