Order Entry
Austria
ContactUsLinkComponent
2H-Perfluoro-5-methyl-3,6-dioxanonane 97%
2H-Perfluoro-5-methyl-3,6-dioxanonane 97%
Catalog # L16954.06
CAS Number:  
2H-Perfluoro-5-methyl-3,6-dioxanonane 97%
Catalog # L16954.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:3330-14-1
 :
Formula:C₈HF₁₇O₂
MDL Number:MFCD00054714
 :452.07 g/mol
Boiling Pt:103…105 °C
Melting Pt:–122 °C
Density:1.659
Storage Temperature:Ambient