

(S)-(-)-1-(p-Tolyl)ethylamine ≥98%, ee 99%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27298-98-2
:
Formula:C₉H₁₃N
MDL Number:MFCD00145246
:135.21 g/mol
Boiling Pt:204 °C
Melting Pt:< –20 °C
Density:0.94
Flash Pt:82 °C (179 °F)
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together