

2-Trifluoromethyl-2-propanol 98%
Catalog # L10888.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:507-52-8
:
Formula:C₄H₇F₃O
MDL Number:MFCD00014391
:128.09 g/mol
Boiling Pt:80…81 °C
Density:1.170
Flash Pt:15 °C (59 °F)
Storage Temperature:Ambient
EINECS:208-075-3
ADR:3,II
:GHS02
:GHS07