Order Entry
Austria
ContactUsLinkComponent
Ethidium bromide 98% for molecular biology
Toxic
Health
Ethidium bromide 98% for molecular biology
Catalog # J67270.14
CAS Number:  
Toxic
Health
Ethidium bromide 98% for molecular biology
Catalog # J67270.14
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:∼ 261 °C (dec.)
EINECS:214-984-6
ADR:6.1,I
 :394.31 g/mol
UN:2811
 :GHS06
 :GHS08
Storage Temperature:Ambient
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724