2'-O-Methyladenosine 99%
Catalog # J64922.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Environmentally Preferable:
Specifications
About this item
Synonyms: (2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)-4-methoxytetrahydrofuran-3-ol
:
:281.27 g/mol
CAS Number:2140-79-6
Formula:C₁₁H₁₅N₅O₄
MDL Number:MFCD00056002