Order Entry
Austria
ContactUsLinkComponent
Borate buffers, 0.5 M solutions, pH 8,0 to 9,5
Borate buffers, 0.5 M solutions, pH 8,0 to 9,5
Certificates
Borate buffers, 0.5 M solutions, pH 8,0 to 9,5
Certificates

Specifications

  • CAS No.:
    1330-43-4
  • MDL No.:
    MFCD00163147
  • Physical form:
    Liquid

Specifications

Frequently Bought Together