Order Entry
Austria
ContactUsLinkComponent
IEF Sample buffer (4X) pH 3 to 7 with lysine
IEF Sample buffer (4X) pH 3 to 7 with lysine
Catalog # J63633.AC
CAS Number:  
IEF Sample buffer (4X) pH 3 to 7 with lysine
Catalog # J63633.AC
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Description:
    IEF Sample Buffer (4X) with Lysine
  • Physical form:
    Liquid
  • pH value:
    3 - 7

Specifications

About this item

Physical Form: Liquid