Order Entry
Austria
ContactUsLinkComponent
Tween 20 washing buffer, 1% in PBS (10X)
Tween 20 washing buffer, 1% in PBS (10X)
Certificates
Tween 20 washing buffer, 1% in PBS (10X)
Certificates

Specifications

  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Viscous liquid

Specifications

Related Information

Frequently Bought Together