Order Entry
Austria
ContactUsLinkComponent
Phosphate-citrate Buffer (5X)
Phosphate-citrate Buffer (5X)
Catalog # J62982.AE
CAS Number:  
Phosphate-citrate Buffer (5X)
Catalog # J62982.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Phosphate-citrate buffer (5X)
  • MDL No.:
    MFCD00131859
  • pH value:
    2,5

Specifications

About this item

Phosphate-Citrate Buffer (5X) is used as a substrate buffer in immunoassay procedures. It is used in SDS PAGE electrophoresis.

Contains 1M sodium phosphate and 0.5M sodium citrate.