Order Entry
Austria
ContactUsLinkComponent
 
TRIS-buffered saline (TBS, 10X) pH 7,4, for Western blot
 
TRIS-buffered saline (TBS, 10X) pH 7,4, for Western blot
 

 

  • CAS No.:
    77-86-1
  • MDL No.:
    MFCD00004679
  • Physical form:
    Liquid

 

Frequently Bought Together