Order Entry
Austria
ContactUsLinkComponent
TE buffer, pH 8,1, RNAse free
TE buffer, pH 8,1, RNAse free
Certificates
TE buffer, pH 8,1, RNAse free
Certificates

Specifications

  • MDL No.:
    MFCD00236359
  • Storage temperature:
    Ambient temperature
  • Physical form:
    Liquid

Specifications

Related Information

Frequently Bought Together