Order Entry
Austria
ContactUsLinkComponent
TRIS-buffered saline (TBS, 10X), with 1% Triton® X-100
TRIS-buffered saline (TBS, 10X), with 1% Triton® X-100
Catalog # J62533.AP
CAS Number:  
TRIS-buffered saline (TBS, 10X), with 1% Triton® X-100
Catalog # J62533.AP
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    TRIS-buffered saline (TBS, 10X), with 1% Triton® X-100
  • MDL No.:
    MFCD00132476
  • Physical form:
    Liquid

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00132476