DNA marker, broad range
Supplier: Thermo Fisher Scientific
Certificates
About this item
From 250 to 5000 base pairs: 5 bands from 125 to 1000 bp; four bands from 1500 to 5000 bp. Recommended load volume 20 µl.
Physical Form: Liquid
Specifications
- Form:Liquid